Product Details

SNP ID
rs17720864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:135187715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCCGTATATTACATTAATATCTC[C/T]TGTATTCGGATACTTTAGAGAGACT
Phenotype
MIM: 189990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYB PubMed Links

Gene Details

Gene
MYB
Gene Name
MYB proto-oncogene, transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130172.1 Intron NP_001123644.1
NM_001130173.1 Intron NP_001123645.1
NM_001161656.1 Intron NP_001155128.1
NM_001161657.1 Intron NP_001155129.1
NM_001161658.1 Intron NP_001155130.1
NM_001161659.1 Intron NP_001155131.1
NM_001161660.1 Intron NP_001155132.1
NM_005375.3 Intron NP_005366.2

View Full Product Details