Product Details

SNP ID
rs17812468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73363474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGGGATTTCTGGCTCCTACTCC[C/G]GCAGGCCGCTTGGGTGACTGTCCTT
Phenotype
MIM: 611111 MIM: 611687 MIM: 611689
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPPA5 PubMed Links

Gene Details

Gene
DPPA5
Gene Name
developmental pluripotency associated 5
There are no transcripts associated with this gene.

Gene
KHDC3L
Gene Name
KH domain containing 3 like, subcortical maternal complex member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017361.2 Intron NP_001017361.1
Gene
OOEP
Gene Name
oocyte expressed protein
There are no transcripts associated with this gene.

Gene
OOEP-AS1
Gene Name
OOEP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details