Product Details

SNP ID
rs17002176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:26466705 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGGCACTTTGAGAGTCTTAGCC[A/G]CAGTGTAATTGGGTCTGACCACACG
Phenotype
MIM: 616020 MIM: 616021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYYR1 PubMed Links

Gene Details

Gene
CYYR1
Gene Name
cysteine and tyrosine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320768.1 2725 UTR 3 NP_001307697.1
NM_052954.3 2725 UTR 3 NP_443186.2
XM_011529450.2 2725 Intron XP_011527752.1
Gene
CYYR1-AS1
Gene Name
cysteine and tyrosine rich 1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details