Product Details

SNP ID
rs11550580
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:152032740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTAGGCCACCAATCAGATTAAG[A/G]AATTCTGAGAAATCTAGCTGACCAT
Phenotype
MIM: 603114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A11 PubMed Links

Gene Details

Gene
S100A11
Gene Name
S100 calcium binding protein A11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005620.1 Intron NP_005611.1

View Full Product Details