Product Details

SNP ID
rs12124782
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:32262867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAGAAAGAAGTGGAGGAAGGG[A/G]GAAAAAAAAAAAAAAAAAAACAGAT
Phenotype
MIM: 153390
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LCK PubMed Links

Gene Details

Gene
LCK
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042771.2 Intron NP_001036236.1
NM_005356.4 Intron NP_005347.3
XM_011541453.2 Intron XP_011539755.1
XM_011541454.2 Intron XP_011539756.1
XM_011541455.2 Intron XP_011539757.1

View Full Product Details