Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281956.1 | 1919 | Intron | NP_001268885.1 | ||
NM_052896.4 | 1919 | Intron | NP_443128.2 | ||
XM_017000185.1 | 1919 | Intron | XP_016855674.1 | ||
XM_017000186.1 | 1919 | Intron | XP_016855675.1 | ||
XM_017000187.1 | 1919 | Intron | XP_016855676.1 | ||
XM_017000188.1 | 1919 | Intron | XP_016855677.1 | ||
XM_017000189.1 | 1919 | Intron | XP_016855678.1 | ||
XM_017000190.1 | 1919 | Intron | XP_016855679.1 | ||
XM_017000191.1 | 1919 | Intron | XP_016855680.1 | ||
XM_017000192.1 | 1919 | Intron | XP_016855681.1 | ||
XM_017000193.1 | 1919 | Intron | XP_016855682.1 | ||
XM_017000194.1 | 1919 | Intron | XP_016855683.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_145205.5 | 1919 | Missense Mutation | AAA,AGA | K59R | NP_660206.2 |