Product Details

SNP ID
rs11554216
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:35715716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCGGTTGTCTAGGATGGTGTC[A/G]AAGCTCTCCTGCAGTCGGGCCTGCT
Phenotype
MIM: 605434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf216 PubMed Links

Gene Details

Gene
C1orf216
Gene Name
chromosome 1 open reading frame 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152374.1 1094 Silent Mutation TTC,TTT F202F NP_689587.1
Gene
CLSPN
Gene Name
claspin
There are no transcripts associated with this gene.

View Full Product Details