Product Details
- SNP ID
-
rs3915536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:16719151 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCCACTTAGACTAACGAGGGTCA[C/T]CCATCAGAGTAACCCACTCCCCGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ESPNP
PubMed Links
Gene Details
- Gene
- ESPNP
- Gene Name
- espin pseudogene
There are no transcripts associated with this gene.
View Full Product Details