Product Details

SNP ID
rs6722332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202880604 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAATCTGAATTCAAACCAATACC[C/T]TTAAAGAGAAATGTAAAACTCAGTT
Phenotype
MIM: 616620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICA1L PubMed Links

Gene Details

Gene
ICA1L
Gene Name
islet cell autoantigen 1 like
There are no transcripts associated with this gene.

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 2155 UTR 3 NP_060726.3
XM_011511469.2 2155 UTR 3 XP_011509771.1

View Full Product Details