Product Details

SNP ID
rs11557343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227330654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGGGCAGCATTTCCTTCTCCC[A/G]CTGCTGCTGAGATGGCAGAAATTAG
Phenotype
MIM: 614785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC654841 PubMed Links

Gene Details

Gene
LOC654841
Gene Name
uncharacterized LOC654841
There are no transcripts associated with this gene.

Gene
MFF
Gene Name
mitochondrial fission factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277061.1 580 Intron NP_001263990.1
NM_001277062.1 580 Intron NP_001263991.1
NM_001277063.1 580 Intron NP_001263992.1
NM_001277064.1 580 Intron NP_001263993.1
NM_001277065.1 580 Intron NP_001263994.1
NM_001277066.1 580 Intron NP_001263995.1
NM_001277067.1 580 Intron NP_001263996.1
NM_001277068.1 580 Intron NP_001263997.1
NM_020194.5 580 Intron NP_064579.3
XM_005246681.3 580 Intron XP_005246738.1
XM_005246682.1 580 Intron XP_005246739.1
XM_005246683.1 580 Intron XP_005246740.1
XM_005246684.1 580 Intron XP_005246741.1
XM_005246685.3 580 Intron XP_005246742.1
XM_005246686.3 580 Intron XP_005246743.1
XM_005246687.3 580 Intron XP_005246744.1
XM_005246688.1 580 Intron XP_005246745.1
XM_006712637.1 580 Intron XP_006712700.1
XM_006712638.1 580 Intron XP_006712701.1
XM_006712639.2 580 Intron XP_006712702.1
XM_011511500.1 580 Intron XP_011509802.1
XM_011511501.1 580 Intron XP_011509803.1
XM_017004512.1 580 Missense Mutation ACT,GCT T23A XP_016860001.1
XM_017004513.1 580 Intron XP_016860002.1
XM_017004514.1 580 Intron XP_016860003.1
XM_017004515.1 580 Intron XP_016860004.1

View Full Product Details