Product Details

SNP ID
rs7760210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:166368230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGACTAGTGGCTCTCAAATTG[A/G]ACAGCAAGGCTTTGGAGGAAATATT
Phenotype
MIM: 614738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPC1 PubMed Links

Gene Details

Gene
MPC1
Gene Name
mitochondrial pyruvate carrier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270879.1 Intron NP_001257808.1
NM_016098.3 Intron NP_057182.1
XM_011535894.1 Intron XP_011534196.1
XM_011535895.1 Intron XP_011534197.1

View Full Product Details