Product Details

SNP ID
rs6941499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41267575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGTAGGCAGTGCAGCTCAGGA[C/T]GAAGGTAGTAAAGATTAAAAAAAAC
Phenotype
MIM: 605085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TREM1 PubMed Links

Gene Details

Gene
TREM1
Gene Name
triggering receptor expressed on myeloid cells 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242589.2 5450 Intron NP_001229518.1
NM_001242590.2 5450 Intron NP_001229519.1
NM_018643.4 5450 Intron NP_061113.1
XM_006715117.3 5450 Intron XP_006715180.1
XM_011514696.1 5450 UTR 3 XP_011512998.1
XM_011514697.1 5450 Intron XP_011512999.1
XM_017010956.1 5450 UTR 3 XP_016866445.1
XM_017010957.1 5450 UTR 3 XP_016866446.1

View Full Product Details