Product Details

SNP ID
rs11542739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125238790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCAAATCAGAATCTTCCAAC[A/T]CTTTCTGGACGGGCTTCATAGTAGA
Phenotype
MIM: 138120 MIM: 605962
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPA5 PubMed Links

Gene Details

Gene
HSPA5
Gene Name
heat shock protein family A (Hsp70) member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005347.4 1295 Missense Mutation GAG,GTG E345V NP_005338.1
Gene
RABEPK
Gene Name
Rab9 effector protein with kelch motifs
There are no transcripts associated with this gene.

View Full Product Details