Product Details

SNP ID
rs7972547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25197861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTTAAAGAATGAAGTAGTTAAT[A/G]TAGTGGGTTAAATACCCTCATCTAA
Phenotype
MIM: 616906 MIM: 190070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASC1 PubMed Links

Gene Details

Gene
CASC1
Gene Name
cancer susceptibility candidate 1
There are no transcripts associated with this gene.

Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
There are no transcripts associated with this gene.

Gene
LYRM5
Gene Name
LYR motif containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001660.2 Intron NP_001001660.2
XM_005253319.4 Intron XP_005253376.1
XM_005253320.4 Intron XP_005253377.1
XM_017018850.1 Intron XP_016874339.1

View Full Product Details