Product Details
- SNP ID
-
hCV43492272
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:112797531 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGATTTAGCATGTCGCTCCCATT[C/G]CCTGCTAACCTCAGACAATTCCAAG
- Phenotype
-
MIM: 612159
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPH3A
PubMed Links
Gene Details
- Gene
- RPH3A
- Gene Name
- rabphilin 3A
There are no transcripts associated with this gene.
View Full Product Details