Product Details

SNP ID
rs11545980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99501745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGGCCTGGCGCCCGTCAACTTC[C/T]GCGACGAAGAAAAAAAGAGCGACGA
Phenotype
MIM: 604678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00449 PubMed Links

Gene Details

Gene
LINC00449
Gene Name
long intergenic non-protein coding RNA 449
There are no transcripts associated with this gene.

Gene
LINC01232
Gene Name
long intergenic non-protein coding RNA 1232
There are no transcripts associated with this gene.

Gene
TM9SF2
Gene Name
transmembrane 9 superfamily member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004800.2 372 Missense Mutation CGC,TGC R47C NP_004791.1

View Full Product Details