Product Details
- SNP ID
-
rs8025846
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:40596124 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAGTGGTTCCCAAATTTTCTTGAT[A/G]CATGGCACCCTGAGATCAAAGAAAT
- Phenotype
-
MIM: 609173
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KNL1
PubMed Links
Gene Details
- Gene
- KNL1
- Gene Name
- kinetochore scaffold 1
View Full Product Details