Product Details

SNP ID
rs5745909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75349373 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCACCAGGTGTGATGAACATAGTC[A/G]CGGGCTCCACACCTGACTCTCTTAG
Phenotype
MIM: 616701 MIM: 154580 MIM: 608844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMMD4 PubMed Links

Gene Details

Gene
COMMD4
Gene Name
COMM domain containing 4
There are no transcripts associated with this gene.

Gene
MAN2C1
Gene Name
mannosidase alpha class 2C member 1
There are no transcripts associated with this gene.

Gene
MIR631
Gene Name
microRNA 631
There are no transcripts associated with this gene.

Gene
NEIL1
Gene Name
nei like DNA glycosylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256552.1 Intron NP_001243481.1
NM_024608.3 Intron NP_078884.2
XM_005254659.4 Intron XP_005254716.1
XM_006720677.3 Intron XP_006720740.1
XM_006720678.3 Intron XP_006720741.1
XM_006720679.3 Intron XP_006720742.1
XM_006720680.1 Intron XP_006720743.1
XM_006720681.1 Intron XP_006720744.1
XM_011522001.2 Intron XP_011520303.2
XM_011522002.1 Intron XP_011520304.1
XM_011522003.2 Intron XP_011520305.1
XM_011522004.2 Intron XP_011520306.1
XM_011522005.1 Intron XP_011520307.1
XM_011522006.1 Intron XP_011520308.1
XM_011522007.1 Intron XP_011520309.1
XM_011522008.1 Intron XP_011520310.1
XM_017022566.1 Intron XP_016878055.1
XM_017022567.1 Intron XP_016878056.1
XM_017022568.1 Intron XP_016878057.1

View Full Product Details