Product Details

SNP ID
rs11547106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90271395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGGCTCTGGAAATACCTCAAA[A/G]CTGCTCCCTGCAGGCCACTCTGTAT
Phenotype
MIM: 602293 MIM: 616718
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIB1 PubMed Links

Gene Details

Gene
CIB1
Gene Name
calcium and integrin binding 1
There are no transcripts associated with this gene.

Gene
LOC400446
Gene Name
PERP, TP53 apoptosis effector pseudogene
There are no transcripts associated with this gene.

Gene
NGRN
Gene Name
neugrin, neurite outgrowth associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033088.1 Intron NP_001028260.2

View Full Product Details