Product Details

SNP ID
rs11544150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81889242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGGCATCTGCCTTCCGCTCAA[A/G]GTGCACGTCTGCTGTTCCTAAGCTG
Phenotype
MIM: 604171 MIM: 614534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005782.3 484 Missense Mutation CTT,TTT L160F NP_005773.3
Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
There are no transcripts associated with this gene.

View Full Product Details