Product Details

SNP ID
rs4441330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18659407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGACTGGTTTAAACCCTTTATC[C/T]GACTTCTTGATGAATGCTCTGGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXO3B PubMed Links

Gene Details

Gene
FOXO3B
Gene Name
forkhead box O3B pseudogene
There are no transcripts associated with this gene.

Gene
ZNF286B
Gene Name
zinc finger protein 286B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145045.1 Intron NP_001138517.1

View Full Product Details