Product Details

SNP ID
rs11547455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42316866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAATGAATCTAAAGTGCGGGGG[A/G]ACATCGGCAGGTCAATGGTATTGCT
Phenotype
MIM: 102582 MIM: 601511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAT3 PubMed Links

Gene Details

Gene
STAT3
Gene Name
signal transducer and activator of transcription 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003150.3 2406 Missense Mutation TCC,TTC S726F NP_003141.2
NM_139276.2 2406 Missense Mutation TCC,TTC S727F NP_644805.1
NM_213662.1 2406 Intron NP_998827.1
XM_005257616.3 2406 Missense Mutation TCC,TTC S726F XP_005257673.2
XM_005257617.3 2406 Missense Mutation TCC,TTC S727F XP_005257674.2
XM_011525145.2 2406 Missense Mutation TCC,TTC S727F XP_011523447.1
XM_011525146.2 2406 Intron XP_011523448.1
XM_017024972.1 2406 Intron XP_016880461.1
XM_017024973.1 2406 Intron XP_016880462.1
XM_017024974.1 2406 Intron XP_016880463.1
XM_017024975.1 2406 Missense Mutation TCC,TTC S726F XP_016880464.1
XM_017024976.1 2406 Intron XP_016880465.1
Gene
STAT5A
Gene Name
signal transducer and activator of transcription 5A
There are no transcripts associated with this gene.

View Full Product Details