Product Details

SNP ID
rs4988358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61683219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAAGTATGCCACTTATTCAAT[G/T]TACAAATTATTACTTACAACAGAGT
Phenotype
MIM: 605882
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BRIP1 PubMed Links

Gene Details

Gene
BRIP1
Gene Name
BRCA1 interacting protein C-terminal helicase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032043.2 4624 UTR 3 NP_114432.2
XM_011525332.2 4624 UTR 3 XP_011523634.1
XM_011525333.2 4624 UTR 3 XP_011523635.1
XM_011525334.2 4624 UTR 3 XP_011523636.1
XM_011525335.2 4624 UTR 3 XP_011523637.1
XM_011525336.2 4624 UTR 3 XP_011523638.1
XM_011525337.2 4624 UTR 3 XP_011523639.1
XM_011525338.2 4624 UTR 3 XP_011523640.1
XM_011525339.2 4624 Intron XP_011523641.1
XM_011525340.2 4624 Intron XP_011523642.1
XM_011525341.2 4624 Intron XP_011523643.1
XM_017025200.1 4624 UTR 3 XP_016880689.1
XM_017025201.1 4624 UTR 3 XP_016880690.1
XM_017025202.1 4624 UTR 3 XP_016880691.1
XM_017025203.1 4624 UTR 3 XP_016880692.1

View Full Product Details