Product Details
- SNP ID
-
rs7212400
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:64548898 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGTTTATAGCAGTGAATTCCATGT[A/G]TACTATAAAACATAACTTGCAAATC
- Phenotype
-
MIM: 605532
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SMURF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74375784] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMURF2
- Gene Name
- SMAD specific E3 ubiquitin protein ligase 2
View Full Product Details