Product Details

SNP ID
rs8083916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46114710 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTCTCCCAGGAAGAAGGCCTTC[A/C]AGCTGGGAGCTGGGAGCAAAGAAAG
Phenotype
MIM: 608775
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAUS1 PubMed Links

Gene Details

Gene
HAUS1
Gene Name
HAUS augmin like complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138443.3 Intron NP_612452.1

View Full Product Details