Product Details

SNP ID
rs1058673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42910824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCATGACCTCCCTGGGGTTTAA[A/G]TTGCTGCTGGAGATGGAGGGCTTGG
Phenotype
MIM: 176395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSG6 PubMed Links

Gene Details

Gene
PSG6
Gene Name
pregnancy specific beta-1-glycoprotein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031850.3 594 Silent Mutation NP_001027020.1
NM_002782.4 594 Intron NP_002773.1

View Full Product Details