Product Details

SNP ID
rs11550999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35868748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTGCGCGCGCAGCCCGCCATC[A/G]GGAGCCTGGCCGGTGGGAGCCCCGG
Phenotype
MIM: 104775 MIM: 607762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APLP1 PubMed Links

Gene Details

Gene
APLP1
Gene Name
amyloid beta precursor like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024807.2 304 Missense Mutation AGG,GGG R38G NP_001019978.1
NM_005166.4 304 Missense Mutation AGG,GGG R38G NP_005157.1
XM_017026737.1 304 Missense Mutation AGG,GGG R38G XP_016882226.1
XM_017026738.1 304 Missense Mutation AGG,GGG R38G XP_016882227.1
Gene
KIRREL2
Gene Name
kin of IRRE like 2 (Drosophila)
There are no transcripts associated with this gene.

View Full Product Details