Product Details

SNP ID
hCV44008707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43528996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCTGGAAGACAGAGAAAAGGG[G/T]CCTCTGAGGCTTGTTTGGTGCCCTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB6 PubMed Links

Gene Details

Gene
EFCAB6
Gene Name
EF-hand calcium binding domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022785.3 Intron NP_073622.2
NM_198856.2 Intron NP_942153.1
Gene
EFCAB6-AS1
Gene Name
EFCAB6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details