Product Details

SNP ID
rs8138476
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43961284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATTTGTAAGAATCGCTTGGTGC[C/T]ACACTAGACCTTCTGTGTCCATCTC
Phenotype
MIM: 612058
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SAMM50 PubMed Links
Additional Information
For this assay, SNP(s) [rs201208438] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SAMM50
Gene Name
SAMM50 sorting and assembly machinery component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015380.4 Intron NP_056195.3

View Full Product Details