Product Details
- SNP ID
-
rs6525583
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:72159172 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCAAATTTTTCTGCTGTGTTTCC[C/G]TTTTCTCCAGAAATTCACCATAAGC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ44635
PubMed Links
Gene Details
- Gene
- FLJ44635
- Gene Name
- TPT1-like protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207422.2 |
|
Intron |
|
|
NP_997305.2 |
- Gene
- NHSL2
- Gene Name
- NHS like 2
There are no transcripts associated with this gene.
View Full Product Details