Product Details

SNP ID
rs7654199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75954073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATCACCACACAAAATTATTGG[G/T]TTTTGAAAAGTACCCACGCCTGGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SDAD1 PubMed Links

Gene Details

Gene
SDAD1
Gene Name
SDA1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288983.1 Intron NP_001275912.1
NM_001288984.1 Intron NP_001275913.1
NM_018115.3 Intron NP_060585.2
XM_005263102.4 Intron XP_005263159.1
XM_005263105.4 Intron XP_005263162.1
XM_017008354.1 Intron XP_016863843.1
XM_017008355.1 Intron XP_016863844.1

View Full Product Details