Product Details

SNP ID
rs11542220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69085137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCCGGACTTCAGGGACATTTC[C/G]AAATGGAACAACCGCGTAGTGAGCA
Phenotype
MIM: 605709 MIM: 603172
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL6IP5 PubMed Links

Gene Details

Gene
ARL6IP5
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006407.3 199 Silent Mutation TCC,TCG S30S NP_006398.1
Gene
UBA3
Gene Name
ubiquitin like modifier activating enzyme 3
There are no transcripts associated with this gene.

View Full Product Details