Product Details
- SNP ID
-
rs10050544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179691587 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATGAGTCAAATTGCCTGATCA[C/T]GAAAGGGAGAATGAAGCAGATGTCA
- Phenotype
-
MIM: 114217
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CANX
PubMed Links
Gene Details
- Gene
- CANX
- Gene Name
- calnexin
- Gene
- CBY3
- Gene Name
- chibby family member 3
There are no transcripts associated with this gene.
View Full Product Details