Product Details

SNP ID
rs7150054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:72929423 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGTTAAATCTCAGATTTGCTGC[C/T]GCGCGTGGTGGCTCACGCCTGTCAT
Phenotype
MIM: 616372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF4 PubMed Links

Gene Details

Gene
DCAF4
Gene Name
DDB1 and CUL4 associated factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163508.1 Intron NP_001156980.1
NM_001163509.1 Intron NP_001156981.1
NM_015604.3 Intron NP_056419.2
NM_181340.2 Intron NP_851937.1
NM_181341.2 Intron NP_851938.2
XM_017021201.1 Intron XP_016876690.1
XM_017021202.1 Intron XP_016876691.1
XM_017021203.1 Intron XP_016876692.1
XM_017021204.1 Intron XP_016876693.1
XM_017021205.1 Intron XP_016876694.1
XM_017021206.1 Intron XP_016876695.1
XM_017021207.1 Intron XP_016876696.1
XM_017021208.1 Intron XP_016876697.1
XM_017021209.1 Intron XP_016876698.1
XM_017021210.1 Intron XP_016876699.1
XM_017021211.1 Intron XP_016876700.1
XM_017021212.1 Intron XP_016876701.1
XM_017021213.1 Intron XP_016876702.1
XM_017021214.1 Intron XP_016876703.1
XM_017021215.1 Intron XP_016876704.1

View Full Product Details