Product Details

SNP ID
rs8022786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94645332 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGTTGGAAGTGCAGAGGATGCTTC[C/T]TGGGGAGGTCACTCATGCTTTTAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA13P PubMed Links

Gene Details

Gene
SERPINA13P
Gene Name
serpin family A member 13, pseudogene
There are no transcripts associated with this gene.

View Full Product Details