Product Details

SNP ID
hCV44821604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35463032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATCTTGCCTCTGTGATATAAAA[A/G]GCATATAGAAAGTATTTATTTATTT
Phenotype
MIM: 609689 MIM: 601146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP250 PubMed Links

Gene Details

Gene
CEP250
Gene Name
centrosomal protein 250
There are no transcripts associated with this gene.

Gene
GDF5
Gene Name
growth differentiation factor 5
There are no transcripts associated with this gene.

Gene
MIR1289-1
Gene Name
microRNA 1289-1
There are no transcripts associated with this gene.

View Full Product Details