Product Details
- SNP ID
-
rs7928868
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:121294789 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTTTGTGTTCTTTTTGGTTGCAC[A/G]ATACTATTCTAGGTATTGTATTGAA
- Phenotype
-
MIM: 602286
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SC5D
PubMed Links
Gene Details
- Gene
- SC5D
- Gene Name
- sterol-C5-desaturase
View Full Product Details