Product Details

SNP ID
rs11541683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64284227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGCTGCTCCTGCTGGTGACTGG[A/C]GTCCCACAGGAGGCCTGGGGCCTGG
Phenotype
MIM: 603167 MIM: 605720
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BAD PubMed Links

Gene Details

Gene
BAD
Gene Name
BCL2 associated agonist of cell death
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004322.3 478 Missense Mutation GCC,TCC A48S NP_004313.1
NM_032989.2 478 Missense Mutation GCC,TCC A48S NP_116784.1
Gene
GPR137
Gene Name
G protein-coupled receptor 137
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170726.1 478 Intron NP_001164197.1
NM_001170880.1 478 Intron NP_001164351.1
NM_001170881.1 478 Intron NP_001164352.1
NM_001177358.1 478 Intron NP_001170829.1
NM_020155.3 478 Intron NP_064540.3
XM_005274100.2 478 Intron XP_005274157.1
XM_005274101.2 478 Intron XP_005274158.1
XM_005274102.2 478 Intron XP_005274159.1
XM_005274104.2 478 Intron XP_005274161.1
XM_011545168.2 478 Intron XP_011543470.1
XM_011545169.1 478 Intron XP_011543471.1
XM_011545170.2 478 Intron XP_011543472.1
XM_011545171.2 478 Intron XP_011543473.1
XM_011545172.2 478 Intron XP_011543474.1
XM_017018014.1 478 Intron XP_016873503.1
XM_017018015.1 478 Intron XP_016873504.1
XM_017018016.1 478 Intron XP_016873505.1
Gene
KCNK4
Gene Name
potassium two pore domain channel subfamily K member 4
There are no transcripts associated with this gene.

Gene
KCNK4-TEX40
Gene Name
KCNK4-TEX40 readthrough
There are no transcripts associated with this gene.

View Full Product Details