Product Details

SNP ID
rs6930092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:395349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTATCTAATAGTCTTTAGTGAG[C/T]GACTATTTTGCAGAAAAGGAGTAAT
Phenotype
MIM: 601900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF4 PubMed Links

Gene Details

Gene
IRF4
Gene Name
interferon regulatory factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195286.1 Intron NP_001182215.1
NM_002460.3 Intron NP_002451.2
XM_006715090.2 Intron XP_006715153.1

View Full Product Details