Product Details
- SNP ID
-
hCV45140242
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:125578733 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGCCAGACCACCCCATCTCATGC[C/G]TAGACTACTGTGGTCCCTTCAGCCA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TEX36
PubMed Links
Gene Details
- Gene
- TEX36
- Gene Name
- testis expressed 36
- Gene
- TEX36-AS1
- Gene Name
- TEX36 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details