Product Details
- SNP ID
-
rs11539907
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144789860 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCGGAGACGTCCAGTCCGGCGGG[C/T]AGCAATGAGACCCACTTTGCGGCCA
- Phenotype
-
MIM: 604177
MIM: 194526
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6850
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114247992] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6850
- Gene Name
- microRNA 6850
There are no transcripts associated with this gene.
- Gene
- RPL8
- Gene Name
- ribosomal protein L8
- Gene
- ZNF34
- Gene Name
- zinc finger protein 34
There are no transcripts associated with this gene.
- Gene
- ZNF517
- Gene Name
- zinc finger protein 517
There are no transcripts associated with this gene.
View Full Product Details