Product Details

SNP ID
rs11539900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144789909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGGGGCATCTCTGCGGATGGT[G/T]GAGGGCTTGCCGATGTGCTGGTGGT
Phenotype
MIM: 604177 MIM: 194526
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR6850 PubMed Links

Gene Details

Gene
MIR6850
Gene Name
microRNA 6850
There are no transcripts associated with this gene.

Gene
RPL8
Gene Name
ribosomal protein L8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000973.4 804 Silent Mutation TCA,TCC S223S NP_000964.1
NM_001317771.1 804 Silent Mutation TCA,TCC S223S NP_001304700.1
NM_001317782.1 804 Silent Mutation TCA,TCC S223S NP_001304711.1
NM_033301.2 804 Silent Mutation TCA,TCC S223S NP_150644.1
XM_017013723.1 804 Intron XP_016869212.1
Gene
ZNF34
Gene Name
zinc finger protein 34
There are no transcripts associated with this gene.

Gene
ZNF517
Gene Name
zinc finger protein 517
There are no transcripts associated with this gene.

View Full Product Details