Product Details
- SNP ID
-
rs7026345
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:131504749 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGCTCTTTATTAACTGATTAAT[A/G]TGTGTATGTGTGTGTGTGTGTGTGT
- Phenotype
-
MIM: 607423
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POMT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs564470917] are located under a probe and SNP(s) [rs111397669] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POMT1
- Gene Name
- protein O-mannosyltransferase 1
- Gene
- PRRC2B
- Gene Name
- proline rich coiled-coil 2B
There are no transcripts associated with this gene.
View Full Product Details