Product Details

SNP ID
rs28669111
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:2022627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGGTCCAAAAATAAAGCCTCA[C/T]TAGAGCCCTTTGCGTCCGACCTTCA
Phenotype
MIM: 137163
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GABRD PubMed Links

Gene Details

Gene
GABRD
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000815.4 898 Intron NP_000806.2
XM_011541194.2 898 UTR 5 XP_011539496.1
XM_017000936.1 898 UTR 5 XP_016856425.1
Gene
LOC105378589
Gene Name
uncharacterized LOC105378589
There are no transcripts associated with this gene.

View Full Product Details