Product Details

SNP ID
rs28753776
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:2027588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTCTTGGCAGAATCACCTCCA[C/G]TGTGGCCTGCGACATGGACCTGGCC
Phenotype
MIM: 137163
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GABRD PubMed Links

Gene Details

Gene
GABRD
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000815.4 577 Missense Mutation ACT,AGT T161S NP_000806.2
XM_011541194.2 577 Missense Mutation ACT,AGT T174S XP_011539496.1
XM_017000936.1 577 Missense Mutation ACT,AGT T396S XP_016856425.1

View Full Product Details