Product Details

SNP ID
rs28695703
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:968975 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGGCCAACACACAGAGGATCGAC[C/T]CCAGAAATCTCCTCACATATTTCAG
Phenotype
MIM: 610770 MIM: 615921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL17 PubMed Links

Gene Details

Gene
KLHL17
Gene Name
kelch like family member 17
There are no transcripts associated with this gene.

Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
There are no transcripts associated with this gene.

Gene
PERM1
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
There are no transcripts associated with this gene.

Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160184.1 Intron NP_001153656.1
NM_032129.2 Intron NP_115505.2
XM_006710944.3 Intron XP_006711007.2
XM_011542248.2 Intron XP_011540550.2
XM_017002474.1 Intron XP_016857963.1
XM_017002475.1 Intron XP_016857964.1
XM_017002476.1 Intron XP_016857965.1
XM_017002477.1 Intron XP_016857966.1
XM_017002478.1 Intron XP_016857967.1
XM_017002479.1 Intron XP_016857968.1

View Full Product Details