Product Details

SNP ID
hCV57466883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:978935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCACGGCTGGCTTGAGGGGGG[A/T]CCCAGGCCCCGCCCCCTCGGAGGCC
Phenotype
MIM: 615921
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PERM1 PubMed Links

Gene Details

Gene
PERM1
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291366.1 1813 Missense Mutation ACC,TCC T699S NP_001278295.1
NM_001291367.1 1813 Missense Mutation ACC,TCC T605S NP_001278296.1
Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
There are no transcripts associated with this gene.

View Full Product Details