Product Details

SNP ID
rs11545897
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:159587931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTAGGCCAGGAATATGAACAAG[C/T]CGCTGCTTTGGATCTCTGTCCTCAC
Phenotype
MIM: 104770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APCS PubMed Links

Gene Details

Gene
APCS
Gene Name
amyloid P component, serum
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001639.3 106 Missense Mutation CCG,TCG P4S NP_001630.1

View Full Product Details