Product Details

SNP ID
rs34355066
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:150577050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGACTTGCCTGGCTACTGGCCA[A/C]TTTCCTGTTCTCAACAAGGAAATTA
Phenotype
MIM: 159552
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTSL4-AS1 PubMed Links

Gene Details

Gene
ADAMTSL4-AS1
Gene Name
ADAMTSL4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MCL1
Gene Name
BCL2 family apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197320.1 1338 UTR 3 NP_001184249.1
NM_021960.4 1338 UTR 3 NP_068779.1
NM_182763.2 1338 UTR 3 NP_877495.1

View Full Product Details