Product Details
- SNP ID
-
rs28649482
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:172661726 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTAATAAGAAAAGAGATGCAAAT[A/T]AAAAATAGGGAATACCAATTTTCAT
- Phenotype
-
MIM: 134638
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FASLG
PubMed Links
Gene Details
- Gene
- FASLG
- Gene Name
- Fas ligand
View Full Product Details